Learn More
Medchemexpress LLC Palbociclib hydrochloride | 571189-11-2 | MFCD22420809 | 100.0% | 484.0 g/mol | C24H30ClN7O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY50767C10MG
Encompass_Preferred
Palbociclib hydrochloride is an orally active, selective CDK4 and CDK6 inhibitor used in preclinical research. It exhibits potent anti-proliferative activity and induces cell-cycle arrest in cancer cells, with reported IC50 values of 11 nM (CDK4) and 16 nM (CDK6). Supplied as a solid powder for laboratory use.
Catalog No. 50-002-98732