Learn More
Medchemexpress LLC Palbociclib impurity 20 | 1774898-55-3 | 99.9% | 308.34 | 100 MG

Supplier: Medchemexpress LLC HY42925100MG
Palbociclib impurity 20 is an impurity of Palbociclib. This product is intended for research use only and is not for sale to patients.
- High purity of 99.9%
- Molecular weight is 308.34
- Formula: C14H20N4O4
- CAS number: 1774898-55-3
- Appearance: Solid
- SMILES: O=C(N1CCN(C2=C([N+]([O-])=O)C=CN=C2)CC1)OC(C)(C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.