Learn More
Medchemexpress LLC Palbociclib impurity 20 | 1774898-55-3 | 99.9% | 308.34 | 50 MG

Supplier: Medchemexpress LLC HY4292550MG
Palbociclib impurity 20 is an impurity of Palbociclib (HY-50767). This product is for research use only.
- Solid appearance
- Molecular weight 308.34
- Chemical formula C14H20N4O4
- Store powder at -20°C for 3 years, 4°C for 2 years
- Store in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.