Learn More
Aldevron pALD-HELP 10mg
Supplier:  Aldevron 508210
Encompass_Preferred
10mg dispense of the pALD-HELP helper plasmid as a part of a triple transfection AAV system. 11.6 kb, Kanamycin resistance, license and royalty free for research grade through commercial applications. pALD-HELP removes extraneous DNA, including a Hexon fiber coding sequence, improving its safety profile and manufacturing efficiency.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3876