Learn More
Haemonetics Corporation PALL BLOOD TRANSFUSION FILTER
Supplier:  Haemonetics Corporation SQ40S
Encompass_Preferred
Blood Transfusion Filter, Pall's 40micrometer Rated Polyester Screen Media, Protection from Microaggregates, Clots and Particulate Debris from Stored or Salvaged Blood Components, Low Residual Blood Hold-up Volume, Minimal Blood Trauma
Catalog No. NC9567012
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product