Learn More
Cytiva Pall Emflon PFR membrane in Kleenpak Nova inline style capsule 0.2um
Supplier:  Cytiva NP6PFRP1
Encompass
Pall Emflon PFR membrane in Kleenpak Nova inline style capsule 0.2um 10 in 8000 cm2 EFA 1 5 in sanitary flange connections
Catalog No. 50-005-48522