Learn More
Cytiva Pall Emflon PFR membrane in Mini Kleenpak capsules 0.2um, Box of 3
Supplier:  Cytiva KA02PFRP8
Encompass
Pall Emflon PFR membrane in Mini Kleenpak capsules 0.2um sterilizing grade 280 cm2 EFA 0 5 in Triclover-compatible connections box of 3
Catalog No. 50-005-48521