Learn More
FUJIFILM BIOSCIENCES INC Palladium(II) Acetate, 1 g, 46.5-49.5% Pd
Supplier: FUJIFILM BIOSCIENCES INC 16524701
Palladium(II) acetate is a solid palladium catalyst widely used for carbon-carbon coupling reactions in organic synthesis, including Suzuki-Miyaura and Heck reactions. Supplied with a measured palladium content and packaged under inert gas, this reagent is intended for research and synthetic applications requiring a reliable palladium source.
- Used for carbon-carbon coupling reactions such as Suzuki-Miyaura and Heck.
- Pd content 46.5-49.5% by titration.
- Molecular formula (CH3COO)2Pd; molecular weight 224.51 g/mol.
- Available package sizes: 1 g, 5 g, 25 g.
- Supplied under inert gas to maintain catalytic activity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.