Learn More
Medchemexpress LLC Palladium(II) acetylacetonate | 14024-61-4 | MFCD00000025 | 98.0% | 304.62 g·mol⁻¹ | C10H14O4Pd | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW020952100MG
Encompass_Preferred
Palladium(II) acetylacetonate is a coordination complex used as a catalyst precursor and reagent in organic synthesis and materials chemistry. Supplied as a solid powder for research use, it is typically offered with high reported purity and supporting documentation.
  • Used as a catalyst precursor for Pd-mediated cross-coupling and other Pd-catalyzed transformations.
  • Supplied as a solid powder suitable for laboratory handling and storage.
  • High reported purity (~98.0%) with COA and SDS documentation available.
  • Molecular formula C10H14O4Pd and molecular weight 304.62 g·mol⁻¹.
Catalog No. 50-003-01325