Learn More
Medchemexpress LLC Palladium(II) acetylacetonate | 14024-61-4 | MFCD00000025 | 98.2% | 304.62 | C10H14O4Pd | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0209521G
Encompass_Preferred
Palladium(II) acetylacetonate (Pd(acac)2) is a palladium(II) metal-organic complex used as a catalyst and biochemical research reagent. It is a high-purity solid with formula C10H14O4Pd and molecular weight ≈304.62 g·mol-1; reported purities are near 98%.
  • Used as a catalyst in organic synthesis and cross-coupling reactions.
  • Applicable as a biochemical reagent in research assays.
  • High purity suitable for analytical and preparative work.
  • Typically supplied as a solid for laboratory use.
  • Store protected from light and under inert atmosphere to preserve stability.
Catalog No. 50-002-99575