Learn More
Medchemexpress LLC Palladium(II) acetylacetonate | 14024-61-4 | MFCD00000025 | 98.2% | 304.62 | C10H14O4Pd | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW020952500MG
Encompass_Preferred
Palladium(II) acetylacetonate is an air-stable, light-yellow coordination compound used as a palladium source for catalytic reactions and as a reagent or precursor in organic and inorganic synthesis. It is supplied as a solid with documented purity and downloadable COA and SDS files.
  • Air-stable light-yellow solid.
  • Used as a palladium source for homogeneous catalysis and cross-coupling reactions.
  • High purity (about 98%) with certificate of analysis available.
  • Available in multiple laboratory package sizes for small-scale reactions.
  • Store at 4°C, protect from light, and keep under inert atmosphere.
Catalog No. 50-002-99507