Learn More
Saf T Pak Inc Imperium Half Pallet Insulated Shipping Solution
SDP
Supplier:  Saf T Pak Inc 36057
Encompass_Preferred
Inmark's Imperium family of patent-pending pallet shippers provides the performance and flexibility to solve your bulk distribution challenges. Effectively manage and mitigate risk by tuning to your organization's specific risk profile(s), cost targets and business objectives.  By leveraging patent-pending design and advanced materials, as well as simple, modular, and intuitive design, Imperium family seamlessly integrates into any supply chain, improving efficiency and ensuring compliance.FEATURES: Performs to ISTA 7D Standard: Rigorously designed and developed to ISTA 7D Standard, the Imperium family of pallet shippers ensure performance in the chamber is only exceeded by performance in the field.One Shared Platform - 18 Configurations: The Imperium family incorporates 3 sizes with 18 potential configurations all built on the same platform with shared common parts allowing for built-to-fit payload configurations.
Catalog No. 50-190-4585