Learn More
American Radiolabeled Chemicals Inc PALM AC 2-3H 1LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 19851MC
Encompass_Preferred
Palmitic acid-2-3H 1mCi in ethanol, 1 mCi/ml, 20-30 Ci/mmol, M.W. 256.4, Exclusive
Catalog No. 50-352-970