Learn More
Medchemexpress LLC Palmaturbine | 16176-68-4 | 5 MG

Supplier: Medchemexpress LLC HYN23735MG
Palmaturbine is a natural product isolated from T. sinensis. It is intended for research use only and is supplied as a solid with a purity of ≥99.0%.
- Natural product
- Isolated from T. sinensis
- For research use only
- ≥99.0% purity
- Solid appearance
- Store at 4°C, protect from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.