Learn More
Cayman Chemical Palmitate calcium salt
SDP
Supplier:  Cayman Chemical 100102795 G
Encompass_Preferred
A neutral, lipid soluble form of the free acid; increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol
Catalog No. 50-288-9710