Learn More
Medchemexpress LLC Palmitelaidic Acid | 10030-73-6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN234150MG
Encompass_Preferred
Palmitelaidic acid (9-trans-Hexadecenoic acid) is the trans isomer of palmitoleic acid, a fatty acid abundantly found in serum and tissue.
  • Promotes faster glucose uptake, associated with higher insulin concentration.
  • Increases the phosphorylation of AMPK, up-regulates glucokinase, and down-regulates SREBP-1.
  • Boosts FGF-21 production and stimulates PPARα expression.
  • Reduces body weight gain, alleviates hyperglycemia and hypertriglyceridemia, and enhances insulin sensitivity.
  • Down-regulates mRNA expressions of pro-inflammatory adipocytokine genes (TNFα and resistin) in white adipose tissue and lipogenic genes (SREBP-1, FAS, and SCD-1) in the liver.
Catalog No. 50-004-20252