Learn More
Cayman Chemical PalmItelaIdIc AcId 100mg
SDP
Supplier:  Cayman Chemical 9001798100 MG
Encompass_Preferred
A dietary fatty acid; decreases NO production and levels of soluble E-selectin in isolated HAECs at 20 µM; inhibits Sendai virus-induced hemolysis of isolated human erythrocytes at 49 µM; increased serum levels are associated with higher levels of LDL, but lower levels of triglycerides, fasting insulin, blood pressure, and incidence of diabetes
Catalog No. 50-289-5174