Learn More
Cayman Chemical PalmItelaIdIc AcId-d13 500ug
SDP
Supplier:  Cayman Chemical 27713500 UG
Encompass_Preferred
An internal standard for the quantification of palmitelaidic acid by GC- or LC-MS
Catalog No. 50-294-0423