Learn More
Medchemexpress LLC Palmitic acid-13C | 287100-87-2 | 257.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S91MG
Encompass_Preferred
Palmitic acid-13C is the 13C-labeled version of Palmitic acid. Palmitic acid is a long-chain saturated fatty acid naturally occurring in animals and plants. It has been observed to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This compound is intended for research use only.
  • Utilized as a tracer or an internal standard.
  • Suitable for quantitative analysis via methods such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16929