Learn More
Medchemexpress LLC Palmitic acid-13C2 | 86683-25-2 | 258.41 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S1050MG
Encompass_Preferred
Palmitic acid-13C2 is the 13C-labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Induces expression of glucose-regulated protein 78 (GRP78)
  • Induces expression of CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells
Catalog No. 50-004-16765