Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid | 57-10-3 | 256.42 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYN0830R

Encompass_Preferred

Palmitic acid is a long-chain saturated fatty acid found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is also used to establish a cell steatosis model.

  • Long-chain saturated fatty acid
  • Found in animals and plants
  • Induces expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP)
  • Used to establish cell steatosis models
  • Purity of 99.86%
  • Appears as a white to off-white solid
  • Stable for 3 years at room temperature
  • Stable in solvent for 2 years at -80°C or 1 year at -20°C

Catalog No. 50-003-89216


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.