Learn More
Apexbio Technology LLC Palmitic acid(Synonyms: Hexadecanoic acid, Hexadecylic acid, Cetylic acid, Palmitinic acid, Fatty acid C16:0), 5g, CAS: 21096.
SDP
Supplier:  Apexbio Technology LLC N24565000
Encompass_Preferred
Palmitic acid (CAS 57-10-3) is a saturated long-chain fatty acid naturally isolated from palm tree fruits It is involved biologically as a key constituent of cellular membranes and lipid signaling pathways influencing protein palmitoylation a reversible lipid modification regulating protein localization trafficking and activity Palmitic acid also interacts metabolically with pathways involved in insulin signaling inflammation and lipid metabolism Due to these functional roles it is investigated extensively in biomedical research focusing on metabolic disorders inflammatory responses and cellular signaling mechanisms Store under dry cool and sealed conditions
Catalog No. 50-000-61146