Learn More
Cayman Chemical PalmItIc AcId-d2 1g
SDP
Supplier:  Cayman Chemical 90018761 G
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-289-3353