Learn More
Cayman Chemical PalmItIc AcId-d2 500mg
SDP
Supplier:  Cayman Chemical 9001876500 MG
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-289-3859