Learn More
Medchemexpress LLC Palmitic acid-d2 | 62689-96-7 | 258.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S4100MG
Encompass_Preferred
Palmitic acid-d2 is a deuterium-labeled form of Palmitic acid, which is a long-chain saturated fatty acid naturally found in both animals and plants. This compound has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Deuterium labeled
  • Long-chain saturated fatty acid
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 98.0% (also listed as 99.8%)
Catalog No. 50-004-16259