Learn More
Medchemexpress LLC Palmitic acid-d3 | 75736-53-7 | 259.44 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S550MG
Encompass_Preferred
Palmitic acid-d3 is a deuterium-labeled version of palmitic acid, a long-chain saturated fatty acid found in both animals and plants. This compound is suitable for use as a tracer and an internal standard for quantitative analysis, and deuteration can impact pharmacokinetic and metabolic profiles. It presents as a white to off-white solid.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can induce expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
Catalog No. 50-004-16763