Learn More
Medchemexpress LLC Palmitic acid-d3 (sodium) | 347841-37-6 | 281.42 | 50 MG
Supplier: Medchemexpress LLC HYN0830BS250MG
Palmitic acid-d3 (sodium) is the deuterium-labeled Palmitic acid, intended for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.