Learn More
Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | 287.62 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN0830S2
Palmitic acid-d31 is the deuterium-labeled form of Palmitic acid. Palmitic acid is a long-chain saturated fatty acid found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. Stable heavy isotopes like deuterium are incorporated into drug molecules for use as tracers in drug development, with potential to affect pharmacokinetic and metabolic profiles.
- Deuterium-labeled compound
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Purity: 99.56%
- Appearance: Solid, white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.