Learn More
Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | 287.62 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN0830S2
Encompass_Preferred
Palmitic acid-d31 is the deuterium labeled Palmitic acid, a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It can be used as a tracer and as an internal standard for quantitative analysis.
  • Deuterium labeled palmitic acid.
  • Long-chain saturated fatty acid.
  • Found in both animals and plants.
  • Can induce expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89631