Learn More
Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | 287.62 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S250MG
Encompass_Preferred
Palmitic acid-d31 is the deuterium-labeled Palmitic acid. Palmitic acid is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-16174