Learn More
Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | MFCD00053335 | 99.6% | 287.62 | C16HD31O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S210MG
Encompass_Preferred
Palmitic acid-d31 is a deuterium-labeled (d31) palmitic acid used as a stable isotope standard and internal standard for quantitative mass spectrometry, metabolic tracing, and analytical assays. It is supplied at high purity for use in LC-MS and GC-MS workflows.
  • Deuterium-labeled palmitic acid (d31) for isotopic tracing.
  • High purity suitable for analytical standards and quantitation.
  • Suitable as an internal standard in LC-MS and GC-MS workflows.
  • Common applications include metabolic studies and lipid analysis.
  • Available in multiple pack sizes to support method development.
Catalog No. 50-002-95766