Learn More
Cayman Chemical PalmItIc AcId-d31 500mg
SDP
Supplier:  Cayman Chemical 16497500 MG
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-297-0481