Learn More
Medchemexpress LLC Palmitic acid-d31 sodium | 467235-83-2 | 309.60 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0830BS350MG
Encompass_Preferred
Palmitic acid-d31 (sodium) is a deuterium-labeled palmitic acid primarily used for research purposes. This compound, featuring stable heavy isotopes, acts as a tracer and an internal standard in quantitative analysis methods. Deuteration is known for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-16256