Learn More
Medchemexpress LLC Palmitic acid-d4-1 | 75736-47-9 | 260.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S125MG
Encompass_Preferred
Palmitic acid-d4-1 is a laboratory chemical primarily used for the manufacture of various substances. This product is a solid and exhibits a melting point range of 60-63°C.
  • Stable under recommended storage conditions.
  • For long-term storage as a powder, keep at -20°C for up to 3 years or 4°C for up to 2 years.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Product can be shipped at room temperature if transit time is less than 2 weeks.
Catalog No. 50-004-16260