Learn More
Medchemexpress LLC Palmitic acid-d4-1 | 75736-47-9 | 260.45 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S1250MG
Encompass_Preferred
Palmitic acid-d4-1 is a stable isotope of palmitic acid, identified by the CAS number 75736-47-9. This chemical is supplied as a solid with a melting point range of 60-63°C and a molecular weight of 260.45. It is stable under recommended storage conditions and is intended for use as a laboratory chemical in the manufacture of substances.
  • Solid appearance
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications
  • Melting point 60-63°C
Catalog No. 50-004-16959