Learn More
Medchemexpress LLC Palmitic acid-d4 | 75736-49-1 | MFCD00209772 | 99.9% | 260.45 g/mol | C16H28D4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0830S710MG
Encompass_Preferred
Palmitic acid-d4 is the deuterium-labeled (d4) analogue of palmitic acid used as a tracer and internal standard for quantitative analyses. Supplied as a solid in small milligram pack sizes, it offers high isotopic and chemical purity for analytical workflows. Molecular formula C16H28D4O2; molecular weight 260.45 g/mol. Store sealed and protected from moisture and light; in solvent, keep at -80°C for long-term storage.
  • Deuterium-labeled (d4) internal standard for quantitative analysis.
  • High chemical purity (≈99.9%).
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in small milligram pack sizes, including 10 MG.
  • Solid form that is stable when stored sealed and protected from moisture and light.
Catalog No. 50-002-95989