Learn More
Medchemexpress LLC Palmitic acid N-hydroxysuccinimide | 14464-31-4 | 98.0% | 353.50 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0111345G
Encompass_Preferred
Palmitic acid N-hydroxysuccinimide is utilized for conjugating proteins to create targeted delivery vectors. It functions as a lipophilic electrophile and can facilitate the covalent connection between palmitic acid and ovalbumin. It is for research use only.
  • Can be employed in the synthesis of cetacyl derivatives of amino acids, aminoacyl-trNA, coenzyme A, mercaptoacetic acid, and other amino and thiogenic compounds.
Catalog No. 50-004-52082