Learn More
Cayman Chemical PalmItIc AcId13CC2 labeled 1mg
SDP
Supplier:  Cayman Chemical 289161 MG
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-293-9039