Learn More
Medchemexpress LLC Palmitodiolein | 2190-30-9 | 99.8% | 859.39 | 25 MG

Supplier: Medchemexpress LLC 25MGHY112132
Palmitodiolein (Triglyceride POO) is a triacylglycerol found in vegetable oils. It is used in research and forms an N-nitroso compound after interacting with morpholine.
- Liquid appearance
- Colorless to light yellow
- Ships at room temperature in continental US
- Pure form storage: -20°C for 3 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO (100 mg/mL)
- Various in vivo dissolution protocols available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.