Learn More
Cayman Chemical PalmItoleIc AcId Alkyn 100ug
SDP
Supplier:  Cayman Chemical 25362100 UG
Encompass_Preferred
A ω-alkynylated form of palmitoleic acid used in click chemistry; labels wild-type, but not S209A mutant, Wnt3a in mouse fibroblast L-cells expressing Wnt3a; labels secreted wild-type Wnt3a in conditioned media
Catalog No. 50-293-2875