Learn More
Cayman Chemical PalmItoleIc AcId-d13 10mg
SDP
Supplier:  Cayman Chemical 2771410 MG
Encompass_Preferred
An internal standard for the quantification of palmitoleic acid by GC- or LC-MS
Catalog No. 50-293-9608