Learn More
Medchemexpress LLC Palmitoleoyl chlorid 25mg | 40426-22-0 | 272.85 | C16H29ClO | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11695525MG
Encompass_Preferred
Palmitoleoyl chloride is an acyl chloride derivative of palmitoleic acid used as a reactive synthetic intermediate for preparing esters, amides, and lipid-modified compounds in medicinal chemistry and organic synthesis. It is supplied as a technical reagent for laboratory-scale synthesis and requires anhydrous handling.
  • Acyl chloride functional group useful for acylation and derivatization reactions.
  • Purity 95.0% suitable for research and intermediate synthesis.
  • Available in small laboratory pack sizes for milligram-scale work.
  • Reactive toward moisture; recommended anhydrous handling and storage.
  • Safety and quality documents available (SDS, COA) for handling and verification.
Catalog No. 50-002-28279