Learn More
Cayman Chemical PalmItoleoyl ChlorIde 100mg
SDP
Supplier:  Cayman Chemical 22723100 MG
Encompass_Preferred
A derivative of palmitoleic acid; has been used as an intermediate in the synthesis of a variety of compounds
Catalog No. 50-299-5621