Learn More
Cayman Chemical PalmItoyl EthnolamIde-d4 500UG
SDP
Supplier:  Cayman Chemical 9000551500 UG
Encompass_Preferred
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-4791