Learn More
Cayman Chemical PalmItoyl-L-carnItIn-d3chl 5mg
SDP
Supplier:  Cayman Chemical 265695 MG
Encompass_Preferred
An internal standard for the quantification of palmitoyl-L-carnitine by GC- or LC-MS
Catalog No. 50-293-3610