Learn More
Medchemexpress LLC Palmitoyl tripeptide-38 | 1447824-23-8 | 99.8% | 675.96 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP01045MG
Encompass_Preferred
Palmitoyl tripeptide-38 is a bioactive peptide known for its anti-aging properties. It functions by targeting Reactive Oxygen Species (ROS) and plays a role in immunology/inflammation, metabolic enzyme/protease, and NF-κB pathways.
  • Bioactive peptide with anti-aging effect
  • Targets Reactive Oxygen Species (ROS)
  • Involved in immunology/inflammation pathways
  • Involved in metabolic enzyme/protease pathways
  • Involved in NF-κB pathways
Catalog No. 50-004-26034