Learn More
Cayman Chemical PalmItoyl TrIpeptIde-38acet 1g
SDP
Supplier:  Cayman Chemical 275641 G
Encompass_Preferred
A palmitoylated form of the tripeptide Lys-Met(O2)-Lys-OH; increases the levels of collagen I, collagen II, fibronectin, and hyaluronic acid by 187, 97, 109, and 55%, respectively, produced by isolated human dermal fibroblasts at 6 ppm.
Catalog No. 50-294-1486