Learn More
Medchemexpress LLC Palmitoylcarnitine | 1935-18-8 | MFCD00152152 | 100.0% | 399.61 g/mol | C23H45NO4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12635750MG
Encompass_Preferred
Palmitoylcarnitine is an endogenous acylcarnitine involved in fatty acid transport and metabolism. It is provided as a purified research chemical for biochemical and metabolic studies, including enzyme assays, membrane physiology investigations, and metabolomics.
  • Endogenous metabolite relevant to fatty acid metabolism.
  • High purity (reported 99.97%).
  • Molecular formula C23H45NO4 and molecular weight 399.61 g/mol.
  • Supplied as a 50 mg research sample suitable for laboratory use.
  • Compatible with analytical methods such as mass spectrometry and chromatography.
Catalog No. 50-002-36684