Learn More
Cayman Chemical PalmItoylcholInchlorIde 10mg
SDP
Supplier:  Cayman Chemical 900345610 MG
Encompass_Preferred
An acyl choline; inhibits protein kinase C activity at 100 μM; induces hemolysis in rat erythrocytes; plasma levels are decreased in female patients with ME/CFS
Catalog No. 50-289-6131