Learn More
Cayman Chemical PalmItoylDcarnItInchLRI 10mg
SDP
Supplier:  Cayman Chemical 2655210 MG
Encompass_Preferred
A long-chain acylcarnitine; inhibits carnitine palmitoyltransferase (Ki = 2.1 mM for 14C-palmitoylcarnitine synthesis by erythrocyte membranes)
Catalog No. 50-293-3530